Merck
In vivo uptake of β-amyloid by non-plaque associated microglia.
Current Alzheimer research (2012-01-26)
Cheryl A Hawkes, LeHua Deng, Daniela Fenili, Mark Nitz, JoAnne McLaurin
ABSTRACT
The role of microglia in β-amyloid (Aβ) deposition or clearance in the Alzheimer's disease (AD) brain remains unclear. Previous in vivo studies have focused primarily on the association of microglia with Aβ-positive parenchymal plaques, but have given little consideration to the possible interaction between Aβ and non-plaque associated microglia. Further, it is not known if microglia play a direct role in mediating Aβ uptake following anti-aggregant treatment. We report here the identification of Aβ-positive processes throughout the cortex and hippocampus of TgCRND8 mice expressing the human Swedish (KM670/671NL) and Indiana (V717F) amyloid precursor protein mutations, which localized to ionized calcium binding protein-1-positive resident microglia that were not associated with extracellular plaques. Oral administration of 1-deoxy-1-fluoro-scyllo-inositol, a scyllo-inositol analogue, to TgCRND8 mice improved spatial memory impairments and suppressed amyloid pathology in a dose-dependent manner. Further, treatment with 1-deoxy-1- fluoro-scyllo-inositol significantly increased hippocampal intra-microglial Aβ levels without stimulating microglial proliferation or peripheral macrophage recruitment. These results reveal a novel, beneficial role for non-plaque associated microglia in the regulation of cerebral Aβ levels in a mouse model of AD.
Sigma-Aldrich
scyllo-Inositol, ≥98%