Merck
Darier disease: beware the cyst that smells.
Ophthalmic plastic and reconstructive surgery (2010-05-22)
Divya Mathews, Lakshika P Perera, Luciane D Irion, Fiona Carley
ABSTRACT
A 32-year-old woman presented with cystic eyelid lesions accompanied by foul-smelling discharge. Excision biopsy showed that she had Darier disease, and she was referred to dermatology for further management. Despite clinically indolent-looking lesions, biopsy and histology allowed the diagnosis of this rare condition to be made.
Product Description
Clobetasone butyrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clobetasone butyrate, ≥98%
Clobetasone butyrate for system suitability, European Pharmacopoeia (EP) Reference Standard