Merck
  • A population-based analysis of the class effect of beta-blockers after myocardial infarction.
A population-based analysis of the class effect of beta-blockers after myocardial infarction.
American heart journal (2007-01-24)
Stéphane Rinfret, Michal Abrahamowicz, Jack Tu, Karin Humphries, Mark J Eisenberg, Hugues Richard, Peter C Austin, Louise Pilote
ABSTRACT
Long-term treatment with beta-blockers reduces mortality after acute myocardial infarction (AMI). Whether beta-blockers exert a class effect is unknown. We analyzed mortality after AMI in Canadian patients 65 years or older who were discharged from hospital with a diagnosis of AMI from April 1996 to March 2000. Administrative data from Quebec, Ontario, and British Columbia were merged. We compared patients prescribed with metoprolol, acebutolol, or atenolol within 90 days after discharge. Among 31576 patients, 67% were prescribed with metoprolol, 24% with atenolol, and 9% with acebutolol. Clinical characteristics and proportion of days covered with a beta-blocker prescription were similar across groups. Although controlling for time-dependent covariates representing current use and dosage, as well as for age, sex, congestive heart failure, and several other comorbidities, patients who filled a prescription for acebutolol (hazard ratio 0.71, 95% CI 0.62-0.81) or atenolol (hazard ratio 0.79, 95% CI 0.73-0.87) had significantly lower mortality in comparison with metoprolol. The higher mortality observed in patients receiving metoprolol compared with those receiving atenolol or acebutolol challenges the concept of a class effect of beta-blockers for secondary prevention of AMI.
Product Description
Acebutolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Acebutolol hydrochloride, analytical standard