Merck
  • Cell cycle arrest in endometrial carcinoma cells exposed to gonadotropin-releasing hormone analog.
Cell cycle arrest in endometrial carcinoma cells exposed to gonadotropin-releasing hormone analog.
Gynecologic oncology (1999-06-15)
J W Kim, Y S Lee, B K Kim, D C Park, J M Lee, I K Kim, S E Namkoong
ABSTRACT
Gonadotropin-releasing hormone (GnRH) has been shown to have an inhibitory effect on the growth of several hormone-dependent human tumors. We have treated a human endometrial cancer cell line which expresses GnRH receptor with GnRH analog, D-Trp6-LHRH, in order to study whether there are differences in cell cycle kinetic response. Flow cytometric analysis revealed that cultured carcinoma cells showed a cell cycle arrest at the G1-S transition after treatment with 10 microM D-Trp6-LHRH for 36 h. Western blot analysis showed that the level of p16 protein was obvious following 24 h of D-Trp6-LHRH treatment. These results suggest that the mechanism by which GnRH inhibits the growth of endometrial carcinoma cells may include effects on cell cycle arrest.
Sigma-Aldrich
[D-Trp6]-LH-RH, ≥97% (HPLC), powder