Merck
  • BAFF, a novel ligand of the tumor necrosis factor family, stimulates B cell growth.
BAFF, a novel ligand of the tumor necrosis factor family, stimulates B cell growth.
The Journal of experimental medicine (1999-06-08)
P Schneider, F MacKay, V Steiner, K Hofmann, J L Bodmer, N Holler, C Ambrose, P Lawton, S Bixler, H Acha-Orbea, D Valmori, P Romero, C Werner-Favre, R H Zubler, J L Browning, J Tschopp
ABSTRACT
Members of the tumor necrosis factor (TNF) family induce pleiotropic biological responses, including cell growth, differentiation, and even death. Here we describe a novel member of the TNF family, designated BAFF (for B cell activating factor belonging to the TNF family), which is expressed by T cells and dendritic cells. Human BAFF was mapped to chromosome 13q32-34. Membrane-bound BAFF was processed and secreted through the action of a protease whose specificity matches that of the furin family of proprotein convertases. The expression of BAFF receptor appeared to be restricted to B cells. Both membrane-bound and soluble BAFF induced proliferation of anti-immunoglobulin M-stimulated peripheral blood B lymphocytes. Moreover, increased amounts of immunoglobulins were found in supernatants of germinal center-like B cells costimulated with BAFF. These results suggest that BAFF plays an important role as costimulator of B cell proliferation and function.
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, endotoxin, free
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
0.2 mg
Estimated to ship on 18 March 2025
€701.00
1 mg
Estimated to ship on 18 March 2025
€1,410.00
5 x 1 mg
Estimated to ship on 18 March 2025
€1,900.00
Sigma-Aldrich
BAFF, soluble human, >95% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
0.2 mg
Estimated to ship on 18 March 2025
€701.00
1 mg
Estimated to ship on 18 March 2025
€1,410.00
5 x 1 mg
Estimated to ship on 18 March 2025
€1,900.00