Merck
  • Identification of a novel splice site mutation in the DNAAF4 gene of a Chinese patient with primary ciliary dyskinesia.
Identification of a novel splice site mutation in the DNAAF4 gene of a Chinese patient with primary ciliary dyskinesia.
Asian journal of andrology (2023-05-06)
Yang Xu, Jing Wang, Ji-Hai Liu, Qing-Qiang Gao, Bing Wang, Zhi-Peng Xu
ABSTRACT
Primary ciliary dyskinesia (PCD) is a rare hereditary orphan condition that results in variable phenotypes, including infertility. About 50 gene variants are reported in the scientific literature to cause PCD, and among them, dynein axonemal assembly factor 4 ( DNAAF4 ) has been recently reported. DNAAF4 has been implicated in the preassembly of a multiunit dynein protein essential for the normal function of locomotory cilia as well as flagella. In the current study, a single patient belonging to a Chinese family was recruited, having been diagnosed with PCD and asthenoteratozoospermia. The affected individual was a 32-year-old male from a nonconsanguineous family. He also had abnormal spine structure and spinal cord bends at angles diagnosed with scoliosis. Medical reports, laboratory results, and imaging data were investigated. Whole-exome sequencing, Sanger sequencing, immunofluorescence analysis, hematoxylin-eosin staining, and in silico functional analysis, including protein modeling and docking studies, were used. The results identified DNAAF4 disease-related variants and confirmed their pathogenicity. Genetic analysis through whole-exome sequencing identified two pathogenic biallelic variants in the affected individual. The identified variants were a hemizygous splice site c.784-1G>A and heterozygous 20.1 Kb deletion at the DNAAF4 locus, resulting in a truncated and functionless DNAAF4 protein. Immunofluorescence analysis indicated that the inner dynein arm was not present in the sperm flagellum, and sperm morphological analysis revealed small sperm with twisted and curved flagella or lacking flagella. The current study found novel biallelic variants causing PCD and asthenoteratozoospermia, extending the range of DNAAF4 pathogenic variants in PCD and associated with the etiology of asthenoteratozoospermia. These findings will improve our understanding of the etiology of PCD.
Sigma-Aldrich
Anti-α-Tubulin−FITC antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture