Merck
Reducing Taperin Expression Restores Hearing in Grxcr2 Mutant Mice.
Neuroscience (2022-06-26)
Chang Liu, Na Luo, Bo Zhao
ABSTRACT
Recessive mutations in GRXCR2 cause deafness in both humans and mice. In Grxcr2 null hair cells, the sensory receptors for sound in the inner ear, stereocilia are disorganized. Reducing the expression of taperin, a protein that interacts with GRXCR2 at the base of stereocilia, corrects the morphological defects of stereocilia and restores hearing in Grxcr2 null mice. To further validate this finding, this study generated two novel taperin mutant mouse lines that exhibit progressive hearing loss. Then Grxcr2 null mice were crossed with one of these taperin mutant mice. The following morphological analysis revealed that reducing taperin expression indeed corrected stereocilia morphological abnormalities in Grxcr2 null mice. Functional analysis further confirmed that reducing taperin expression partially restored hearing in Grxcr2 null mice.
Sigma-Aldrich
ANTI-TPRN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GRXCR2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution