Merck
  • Oxaliplatin disrupts nucleolar function through biophysical disintegration.
Oxaliplatin disrupts nucleolar function through biophysical disintegration.
Cell reports (2022-11-10)
H Broder Schmidt, Zane A Jaafar, B Erik Wulff, Jason J Rodencal, Kibeom Hong, Mohammad O Aziz-Zanjani, Peter K Jackson, Manuel D Leonetti, Scott J Dixon, Rajat Rohatgi, Onn Brandman
ABSTRACT
Platinum (Pt) compounds such as oxaliplatin are among the most commonly prescribed anti-cancer drugs. Despite their considerable clinical impact, the molecular basis of platinum cytotoxicity and cancer specificity remain unclear. Here we show that oxaliplatin, a backbone for the treatment of colorectal cancer, causes liquid-liquid demixing of nucleoli at clinically relevant concentrations. Our data suggest that this biophysical defect leads to cell-cycle arrest, shutdown of Pol I-mediated transcription, and ultimately cell death. We propose that instead of targeting a single molecule, oxaliplatin preferentially partitions into nucleoli, where it modifies nucleolar RNA and proteins. This mechanism provides a general approach for drugging the increasing number of cellular processes linked to biomolecular condensates.
Sigma-Aldrich
Anti-SURF6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MKI67 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution