Merck
Regioselective monodeprotection of peracetylated carbohydrates.
Nature protocols (2012-09-08)
Marco Filice, Jose M Guisan, Marco Terreni, Jose M Palomo
ABSTRACT
This protocol describes the regioselective deprotection of single hydroxyls in peracetylated monosaccharides and disaccharides by enzymatic or chemoenzymatic strategies. The introduction of a one-pot enzymatic step by using immobilized biocatalysts obviates the requirement to carry out tedious workups and time-consuming purifications. By using this straightforward protocol, different per-O-acetylated glycopyranosides (mono- or disaccharides, 1-substituted or glycals) can be transformed into a whole set of differentially monodeprotected 1-alcohols, 3-alcohols, 4-alcohols and 6-alcohols in high yields. These tailor-made glycosyl acceptors can then be used for stereoselective glycosylation for oligosaccharide and glycoderivative synthesis. They have been successfully used as building blocks to synthesize tailor-made di- and trisaccharides involved in the structure of lacto-N-neo-tetraose and precursors of the tumor-associated carbohydrate antigen T and the antitumoral drug peracetylated β-naphtyl-lactosamine. We are able to prepare a purified monoprotected carbohydrate in between 1 and 4 d. With this protocol, the small library of monodeprotected products can be synthesized in 1-2 weeks.
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Pricing and availability is not currently available.
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Pricing and availability is not currently available.