Merck
  • A tagged parathyroid hormone derivative as a carrier of antibody cargoes transported by the G protein coupled PTH1 receptor.
A tagged parathyroid hormone derivative as a carrier of antibody cargoes transported by the G protein coupled PTH1 receptor.
Peptides (2014-08-17)
Xavier Charest-Morin, Jean-Philippe Fortin, Robert Lodge, Isabelle Allaeys, Patrice E Poubelle, François Marceau
ABSTRACT
Based on the known fact that the parathyroid hormone (PTH) might be extended at its C-terminus with biotechnological protein cargoes, a vector directing the secretion of PTH1-84 C-terminally fused with the antigenic epitope myc (PTH-myc) was exploited. The functional properties and potential of this analog for imaging PTH1R-expressing cells were examined. The PTH-myc construct was recombinantly produced as a conditioned medium (CM) of transfected HEK 293a cells (typical concentrations of 187nM estimated with ELISAs for PTH). PTH-myc CM induced cyclic AMP formations (10min), with a minor loss of potency relative to authentic PTH1-84, and c-Fos expression (1-3h). Treatment of recipient HEK 293a cells transiently expressing PTH1R with PTH-myc CM (supplemented with a fluorescent monoclonal anti-myc tag antibody, either 4A6 or 9E10) allowed the labeling of endosomal structures positive for Rab5 and/or for β-arrestin1 (microscopy, cytofluorometry). Authentic PTH was inactive in this respect, ruling out a non-specific form of endocytosis like pinocytosis. Using a horseradish peroxidase-conjugated secondary antibody, the endocytosis of the PTH-myc-based antibody complex by endogenous PTH1R was evidenced in MG-63 osteoblastoid cells. The secreted construct PTH-myc represents a bona fide agonist that supports the feasibility of transporting cargoes of considerable molecular weight inside cells using arrestin and Rab5-mediated PTH1R endocytosis. PTH-myc is also transported into cells that express PTH1R at a physiological level. Such tagged peptide hormones may be part of a cancer chemotherapy scheme exploiting a modular cytotoxic secondary antibody and the receptor repertoire expressed in a given tumor.
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse