Merck
  • EphA3 as a target for antibody immunotherapy in acute lymphoblastic leukemia.
EphA3 as a target for antibody immunotherapy in acute lymphoblastic leukemia.
Leukemia (2016-12-07)
S Charmsaz, F Al-Ejeh, T M Yeadon, K J Miller, F M Smith, B W Stringer, A S Moore, F-T Lee, L T Cooper, C Stylianou, G T Yarranton, J Woronicz, A M Scott, M Lackmann, A W Boyd
ABSTRACT
The human EphA3 gene was discovered in a pre-B acute lymphoblastic leukemia (pre-B-ALL) using the EphA3-specific monoclonal antibody (mAb), IIIA4, which binds and activates both human and mouse EphA3. We use two models of human pre-B-ALL to examine EphA3 function, demonstrating effects on pre-B-cell receptor signaling. In therapeutic targeting studies, we demonstrated antitumor effects of the IIIA4 mAb in EphA3-expressing leukemic xenografts and no antitumor effect in the xenografts with no EphA3 expression providing evidence that EphA3 is a functional therapeutic target in pre-B-ALL. Here we show that the therapeutic effect of the anti-EphA3 antibody was greatly enhanced by adding an α-particle-emitting 213Bismuth payload.
Sigma-Aldrich
Anti-phospho-Fyn (pTyr530) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies