Merck
  • Hedgehog signaling in the airway epithelium of patients with chronic obstructive pulmonary disease.
Hedgehog signaling in the airway epithelium of patients with chronic obstructive pulmonary disease.
Scientific reports (2019-03-06)
A Tam, M Hughes, K M McNagny, M Obeidat, T L Hackett, J M Leung, T Shaipanich, D R Dorscheid, G K Singhera, C W T Yang, P D Paré, J C Hogg, D Nickle, D D Sin
ABSTRACT
Genome-wide association studies have linked gene variants of the receptor patched homolog 1 (PTCH1) with chronic obstructive pulmonary disease (COPD). However, its biological role in the disease is unclear. Our objective was to determine the expression pattern and biological role of PTCH1 in the lungs of patients with COPD. Airway epithelial-specific PTCH1 protein expression and epithelial morphology were assessed in lung tissues of control and COPD patients. PTCH1 mRNA expression was measured in bronchial epithelial cells obtained from individuals with and without COPD. The effects of PTCH1 siRNA knockdown on epithelial repair and mucous expression were evaluated using human epithelial cell lines. Ptch1+/- mice were used to assess the effect of decreased PTCH1 on mucous expression and airway epithelial phenotypes. Airway epithelial-specific PTCH1 protein expression was significantly increased in subjects with COPD compared to controls, and its expression was associated with total airway epithelial cell count and thickness. PTCH1 knockdown attenuated wound closure and mucous expression in airway epithelial cell lines. Ptch1+/- mice had reduced mucous expression compared to wildtype mice following mucous induction. PTCH1 protein is up-regulated in COPD airway epithelium and may upregulate mucous expression. PTCH1 provides a novel target to reduce chronic bronchitis in COPD patients.
Sigma-Aldrich
Anti-Protein patched homolog 1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-FOXJ1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL3989, purified immunoglobulin
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture