Merck
  • Tyrosine phosphorylation of a 38-kDa capacitation-associated buffalo (Bubalus bubalis) sperm protein is induced by L-arginine and regulated through a cAMP/PKA-independent pathway.
Tyrosine phosphorylation of a 38-kDa capacitation-associated buffalo (Bubalus bubalis) sperm protein is induced by L-arginine and regulated through a cAMP/PKA-independent pathway.
International journal of andrology (2007-03-16)
S C Roy, S K Atreja
ABSTRACT
The aim of the present study was to determine the effect of L-arginine on nitric oxide (NO*) synthesis, capacitation and protein tyrosine phosphorylation in buffalo spermatozoa. Ejaculated buffalo spermatozoa were capacitated in the absence or presence of heparin, or L-arginine or N(omega)-nitro-L-arginine methyl ester (L-NAME), an inhibitor of nitric oxide synthase (NOS) for 6 h. Capacitating spermatozoa generated NO* both spontaneously and following stimulation with L-arginine and L-NAME quenched such L-arginine-induced NO* production. Immunolocalization of NOS suggested for existence of constitutive NOS in buffalo spermatozoa. L-Arginine (10 mm) was found to be a potent capacitating agent and addition of L-NAME to the incubation media attenuated both L-arginine and heparin-induced capacitation and suggested that NO* is involved in the capacitation of buffalo spermatozoa. Two sperm proteins of M(r) 38 000 (p38) and 20 000 (p20) were tyrosine phosphorylated extensively by both heparin and L-arginine. Of these, the tyrosine phosphorylation of p38 was insensitive to both induction by cAMP agonists as well as inhibition by a protein kinase A (PKA) inhibitor. Further, most of these L-arginine-induced tyrosine phosphorylated proteins were localized to the midpiece and principal piece regions of flagellum of capacitated spermatozoa and suggested that sperm flagellum takes active part during capacitation. These results indicated that L-arginine induces capacitation of buffalo spermatozoa through NO* synthesis and tyrosine phosphorylation of specific sperm proteins involving a pathway independent of cAMP/PKA.
Sigma-Aldrich
Anti-Phosphotyrosine antibody, Mouse monoclonal, clone pT-154, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Phosphotyrosine antibody produced in mouse, clone PT-66, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution