Merck
Immunology. Long live the mature B cell--a baffling mystery resolved.
Science (New York, N.Y.) (2001-09-15)
T J Waldschmidt, R J Noelle
ABSTRACT
What determines whether transitional B cells newly emerged from the bone marrow will differentiate further to become mature, long-lived, circulating B lymphocytes? In a Perspective, Waldschmidt and Noelle discuss new findings showing that the TNF family ligand BAFF and its receptor BAFF-R are crucial for selecting transitional B cells into the mature B cell pool (Thompson et al., Schiemann et al.).
Sigma-Aldrich
BAFF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
BAFF Receptor human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Pricing and availability is not currently available.