
tri-Potassium citrate monohydrate

Synonym(s):
tri-Potassium citrate monohydrate
Empirical Formula (Hill Notation):
C6H5K3O7 · xH2O
CAS Number:
Molecular Weight:
306.39 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing