
Senkirkine

Synonym(s):
12-Hydroxy-4-methyl-4,8-secosenecionan-8,11,16-trione, Renardine
Empirical Formula (Hill Notation):
C19H27NO6
CAS Number:
Molecular Weight:
365.42
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing