![Minoxidil for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/113/866/04a663bc-0b9e-4379-9514-9ae5e0c7be53/640/04a663bc-0b9e-4379-9514-9ae5e0c7be53.png)
Minoxidil for system suitability
![Minoxidil for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/113/866/04a663bc-0b9e-4379-9514-9ae5e0c7be53/640/04a663bc-0b9e-4379-9514-9ae5e0c7be53.png)
Synonym(s):
Minoxidil, 6-(1-Piperidinyl)-2,4-pyrimidinediamine 3-oxide, 6-(1-Piperidinyl)pyrimidine-2,4-diamine 3-oxide
Empirical Formula (Hill Notation):
C9H15N5O
CAS Number:
Molecular Weight:
209.25
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing