
Metobromuron

Synonym(s):
N′-(4-Bromophenyl)-N-methoxy-N-methylurea
Empirical Formula (Hill Notation):
C9H11BrN2O2
CAS Number:
Molecular Weight:
259.10
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing