
Metalaxyl Metabolite CGA 108906

Synonym(s):
(±)-2-[(1-Carboxyethyl)(methoxyacetyl)amino]-3-methylbenzoic acid, N-(2-Carboxy-6-methylphenyl)-N-(methoxyacetyl)-DL-alanine
Empirical Formula (Hill Notation):
C14H17NO6
CAS Number:
Molecular Weight:
295.29
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing