
Menaquinone-4

Synonym(s):
Vitamin K2, Menaquinone 4, Menatetrenone
Empirical Formula (Hill Notation):
C31H40O2
CAS Number:
Molecular Weight:
444.65
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing