![Levodropropizine impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/172/142/1084e5dc-ab9c-488c-a193-dcb1303c83ac/640/1084e5dc-ab9c-488c-a193-dcb1303c83ac.png)
Levodropropizine impurity B
![Levodropropizine impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/172/142/1084e5dc-ab9c-488c-a193-dcb1303c83ac/640/1084e5dc-ab9c-488c-a193-dcb1303c83ac.png)
Synonym(s):
1-Phenylpiperazine, N-Phenyldiethylenediamine
Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing