
KiCqStart® One-Step Probe RT-qPCR ReadyMix™

Synonym(s):
KiCqStart® One Step Probe RT-qPCR Readymix™
Products
Product Category
Brand
Color
Physical Form
Number of Reactions
dNTPs
Hot Start
Sample Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing