
Isopentane Solution

Synonym(s):
2-Methylbutane, Isopentane
Linear Formula:
CH3CH2CH(CH3)2
CAS Number:
Molecular Weight:
72.15
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing