
Cyprodinil Metabolite CGA 304075

Synonym(s):
4-(4-Cyclopropyl-6-methylpyrimidin-2-ylamino)phenol
Empirical Formula (Hill Notation):
C14H15N3O
CAS Number:
Molecular Weight:
241.29
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing