
Cefotaxime sodium

Synonym(s):
Cefotaxime sodium salt, Cefotaxim sodium salt
Empirical Formula (Hill Notation):
C16H16N5NaO7S2
CAS Number:
Molecular Weight:
477.45
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade