
Boc-Lys(Z)-OSu

Synonym(s):
Nα-Boc-Nε-Cbz-L-lysine N-hydroxysuccinimide ester, Nα-Boc-Nε-Z-L-lysine hydroxysuccinimide ester, Nε-Z-Nα-Boc-L-lysine hydroxysuccinimide ester
Empirical Formula (Hill Notation):
C23H31N3O8
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing