
BIS-TRIS

Synonym(s):
2,2-Bis(hydroxymethyl)-2,2′,2″-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane, BIS-Tris, ULTROL® Grade
Empirical Formula (Hill Notation):
C8H19NO5
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Research Area
Quality Segment
Application
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing