
Apotryptophanase from Escherichia coli

Synonym(s):
Tryptophanase from Escherichia coli, L-Tryptophan indole-lyase (deaminating)
CAS Number:
Products
Brand
Biological Source
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing