
Anti-INTS6 antibody produced in rabbit

Synonym(s):
Anti-DBI-1 antibody produced in rabbit, Anti-DICE1 antibody produced in rabbit, Anti-Deleted in cancer 1 antibody produced in rabbit, Anti-Int6 antibody produced in rabbit, Anti-Integrator complex subunit 6 antibody produced in rabbit, Anti-Protein DDX26 antibody produced in rabbit, Anti-Protein deleted in cancer 1 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing