Skip to Content
Merck
Amoxicillin trihydrate for performance verification European Pharmacopoeia (EP) Reference Standard

Amoxicillin trihydrate for performance verification

Amoxicillin trihydrate for performance verification European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Amoxicillin trihydrate
Empirical Formula (Hill Notation):
C16H19N3O5S · 3H2O
CAS Number:
Molecular Weight:
419.45

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1