
5(6)-Carboxytetramethylrhodamine N-succinimidyl ester

Empirical Formula (Hill Notation):
C29H25N3O7
CAS Number:
Molecular Weight:
527.52
Products
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing