Skip to Content
Merck
All Photos(2)

Documents

ABE155

Sigma-Aldrich

Anti-phospho Histone H3.1/H3.2 Antibody (Ser87)

from rabbit, purified by affinity chromatography

Synonym(s):

Alpha-synuclein, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, NACP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

bovine, mouse, human

technique(s)

dot blot: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer87)

Gene Information

human ... SNCA(6622)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. Histone H3.1, H3.2 and H3.3 are all closely related Histone H3 variants, that despite differing by only a few amino acids have been shown to have different functional roles.

Specificity

This antibody recognizes Histone H3.1/H3.2 phosphorylated at Ser87. This antibody demonstrates specificity for phospho Histone H3.1/H3.2 (Ser87), but does not cross react with phospho Histone H3.3, or any other modified and non-modified histones.

Immunogen

Epitope: Phosphorylated at Ser87
KLH-conjugated linear peptide corresponding to human Histone H3.1/H3.2 phosphorylated at Ser87.

Application

Detect Histone H3 using this rabbit polyclonal antibody, Anti-phospho Histone H3.1/H3.2 Antibody (Ser87) validated for use in western blotting & Dot Blot.
Dot Blot (Specificity) Analysis: 1µg/mL of this antibody detected phospho Histone H3.1/H3.2 (Ser87) in a panel of unmodified and modified Histones.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by Western Blotting in Colcemid treated HeLa acid extract.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected phospho Histone H3.1/H3.2 (Ser87) in 10 µg of Colcemid treated HeLa acid extract

Target description

~17 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa acid extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service