GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on SUPELCOWAX 10
CONDITIONS
column
SUPELCOWAX 10, 30 m x 0.32 mm I.D., 0.25 μm (24082)
oven
50°C (1 min), 5 °C/min to 225 °C (5 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 25 cm/sec (set at 200 °C)
injection
1 μL 100:1 split
liner
4 mm I.D., cup style
sample
bacterial methyl ester standards in methyl caproate, 10 mg/mL (47080-U)
Description
Legal Information
null