Merck
Sox2 Deacetylation by Sirt1 Is Involved in Mouse Somatic Reprogramming.
Stem cells (Dayton, Ohio) (2015-05-06)
Wen-Li Mu, Ya-Jun Wang, Peng Xu, De-Long Hao, Xiu-Zhen Liu, Ting-Ting Wang, Feng Chen, Hou-Zao Chen, Xiang Lv, De-Pei Liu
ABSTRACT
Mouse somatic cells can be reprogrammed into induced pluripotent stem cells by defined factors known to regulate pluripotency, including Oct4, Sox2, Klf4, and c-Myc. Together with Oct4, Sox2 plays a major role as a master endogenous pluripotent genes trigger in reprogramming. It has been reported that Sirtuin 1 (Sirt1), a member of the Sirtuin family of NAD(+) -dependent protein deacetylases, is involved in embryonic stem cell antioxidation, differentiation, and individual development. However, as a deacetylation enzyme, whether Sirt1 influences reprogramming through its post-translational modification function remains unknown. In this study, we provide evidence that deacetylation of Sox2 by Sirt1 is required for reprogramming. We found that a low level of Sox2 acetylation could significantly increase reprogramming efficiency. Furthermore, we found that Sox2 can be deacetylated by Sirt1 in an Oct4-mediated manner. Compared with wild-type cells, Sirt1-null mouse embryonic fibroblasts exhibit decreased reprogramming efficiency, and overexpression of Sirt1 rescues this defect. In addition, Sirt1 functions in the regulation of reprogramming through deacetylating Sox2. Taken together, we have identified a new regulatory role of Sirt1 in reprogramming and provided a link between deacetylation events and somatic cell reprogramming. Stem Cells 2015;33:2135-2147.
Sigma-Aldrich
Sirtuin 1 human, recombinant, expressed in E. coli, ≥60% (SDS-PAGE)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Myc
Sigma-Aldrich
MISSION® esiRNA, targeting human MYC
Sigma-Aldrich
SIRT1 human, recombinant, expressed in E. coli, N-terminal histidine tagged, ≥90% (SDS-PAGE), buffered aqueous glycerol solution