Merck
  • Is phenylbutazone a genotoxic carcinogen? A weight-of-evidence assessment.
Is phenylbutazone a genotoxic carcinogen? A weight-of-evidence assessment.
Alternatives to laboratory animals : ATLA (2013-08-27)
Robert D Combes
ABSTRACT
Published in silico, in vitro, in vivo laboratory animal and human data, together with information on biotransformation and data from structure-activity analyses with two decision-tree systems (ACToR and Toxtree), have been used in a weight-of-evidence (WoE) assessment to determine whether phenylbutazone (PBZ) is a genotoxic or a non-genotoxic carcinogen. This was undertaken to facilitate the risk assessment of human exposure to this veterinary drug via the consumption of horsemeat from treated animals. Despite problems with data interpretation at all tiers of the database, it was concluded that PBZ behaves like a genotoxic carcinogen with a threshold dose. This conclusion is based mainly on the results of a definitive rodent bioassay, and on the following observations: a) that PBZ has weak in vitro activity only at high concentrations in some genotoxicity assays, accompanied by high levels of cytotoxicity; b) that it (and a major metabolite) is able to cause sister chromatid exchanges in vivo in rodents; and c) that it can induce cytogenetic effects in vivo in humans. It also takes into account the known and predicted activities of the parent drug, some of its metabolites and two structural analogues, and, importantly, several of the drug's other biochemical effects that are unrelated to toxicity. However, this conclusion is not fully supported by all the evidence, and much of the information is based on old papers. Therefore, more studies are required to establish whether the concentration thresholds seen in vitro would translate to dose thresholds for carcinogenicity, such that a safe dose-level could be defined for the purposes of assessing risk. It was disappointing that a WoE approach to evaluating all of the available hazard data, as is increasingly being advocated to improve the hazard identification paradigm, was unable to provide definitive answers in this case, particularly in view of the large numbers of animals that had been used to provide much of the information.
Sigma-Aldrich
Phenylbutazone
Supelco
Phenylbutazone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Phenylbutazone, European Pharmacopoeia (EP) Reference Standard