Merck
  • Lung neoplasms in rodents after chronic administration of dimethyl hydrogen phosphite.
Lung neoplasms in rodents after chronic administration of dimethyl hydrogen phosphite.
Cancer research (1986-01-01)
J K Dunnick, G A Boorman, J K Haseman, J Langloss, R H Cardy, A G Manus
ABSTRACT
Dimethyl hydrogen phosphite (DMHP), an intermediate in the production of insecticides or herbicides, was administered by p.o. gavage for 2 yr to male Fischer 344/N rats and male and female B6C3F1 mice at doses of 0, 100, or 200 mg/kg and to female Fischer 344/N rats at doses of 0, 50 or 100 mg/kg. Dose related toxicity was seen in the lungs of treated male and female rats. The lung lesions were most prevalent in the high dose male rat group which received a dose twice that given to the high dose female rats. Lung lesions included alveolar epithelial hyperplasia, chemically related pneumonia, alveolar-bronchiolar adenoma, alveolar-bronchiolar carcinoma, and squamous cell carcinoma. DMHP also caused neoplastic and nonneoplastic lesions of the forestomach in male rats; a similar but less pronounced effect was observed in female rats. Nonneoplastic lesions associated with administration of DMHP included mineralization of the cerebellum in male rat and focal calcification of the testis in male mice. Under the conditions of this study, there was clear evidence for carcinogenicity for male rats, equivocal evidence for carcinogenicity in female rats, and no evidence for carcinogenicity in either male or female mice. DMHP caused the highest incidence of lung tumors in the male rat of all chemicals studied to date in the National Cancer Institute-National Toxicology Program Carcinogenesis Testing Program.
Sigma-Aldrich
Dimethyl phosphite, 98%