Merck
  • Increased Calcineurin A Expression Is Associated with a Lower Relapse-Free Survival Rate after Colorectal Cancer Surgery.
Increased Calcineurin A Expression Is Associated with a Lower Relapse-Free Survival Rate after Colorectal Cancer Surgery.
Pathobiology : journal of immunopathology, molecular and cellular biology (2016-06-21)
Hiroaki Niitsu, Takao Hinoi, Kazuhiro Sentani, Shoichiro Mukai, Tomohiro Adachi, Yasufumi Saito, Masashi Miguchi, Masatoshi Kochi, Haruki Sada, Naohide Oue, Wataru Yasui, Hideki Ohdan
ABSTRACT
Increased expression of calcineurin in colorectal cancer (CRC) has been reported. Although the oncogenic function has been suggested, the clinical relevance is still unclear. We herein studied calcineurin expression as a prognostic biomarker in patients receiving curative surgery for stages I-III CRC. In 121 patients with stages I-III CRC treated at Hiroshima University between 1997 and 2003, calcineurin A expression was examined using immunohistochemistry (IHC) staining of surgical specimens. Specimens were considered positive for calcineurin A if any IHC-stained cells were observed within the carcinomatous area, and clinicopathological characteristics and survival outcomes were compared between IHC-positive and -negative groups. Calcineurin A was preferentially expressed in the cytoplasm of cancer cells, and a median of 8% of the cells (range: 0-80%; interquartile range: 0-22.5%) were stained within the carcinomatous areas. Of 121 cases, 81 were determined as IHC positive while 40 were determined to be negative. Positive expression of calcineurin A, as well UICC-TNM stage, was associated with low relapse-free survival (RFS) rates in multivariate analyses (hazard ratio = 2.92; 95% CI: 1.27-7.92; p = 0.010). Increased calcineurin A expression is associated with lower RFS rates and may have clinical value in predicting recurrence.
Sigma-Aldrich
Anti-Calcineurin (α-Subunit) antibody, Mouse monoclonal, clone CN-A1, purified from hybridoma cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal Anti-Calcineurin (β-Subunit) antibody produced in mouse, clone CN-B1, ascites fluid