Merck
Exogenous loading of miRNAs into small extracellular vesicles.
Journal of extracellular vesicles (2021-08-12)
Ricardo C de Abreu, Cristiana V Ramos, Clarissa Becher, Miguel Lino, Carlos Jesus, Paula A da Costa Martins, Patrícia A T Martins, Maria João Moreno, Hugo Fernandes, Lino Ferreira
ABSTRACT
Small extracellular vesicles (sEVs), through their natural ability to interact with biological membranes and exploit endogenous processing pathways to convey biological information, are quintessential for the delivery of therapeutically relevant compounds, such as microRNAs (miRNAs) and proteins. Here, we used a fluorescently-labelled miRNA to quantify the efficiency of different methods to modulate the cargo of sEVs. Our results showed that, compared with electroporation, heat shock, permeation by a detergent-based compound (saponin) or cholesterol-modification of the miRNA, Exo-Fect was the most efficient method with > 50% transfection efficiency. Furthermore, qRT-PCR data showed that, compared with native sEVs, Exo-Fect modulation led to a > 1000-fold upregulation of the miRNA of interest. Importantly, this upregulation was observed for sEVs isolated from multiple sources. The modulated sEVs were able to delivery miR-155-5p into a reporter cell line, confirming the successful delivery of the miRNA to the target cell and, more importantly, its functionality. Finally, we showed that the membrane of Exo-Fect-loaded sEVs was altered compared with native sEVs and that enhanced the internalization of Exo-Fect-loaded sEVs within the target cells and decreased the interaction of those modulated sEVs with lysosomes.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type III-A, ≥85% RNase A basis (SDS-PAGE), 85-140 Kunitz units/mg protein