Merck
  • Myocyte enhancer factor 2D promotes tumorigenicity in malignant glioma cells.
Myocyte enhancer factor 2D promotes tumorigenicity in malignant glioma cells.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-08-04)
Youguang Zhao, Ying Li, Yuan Ma, Songtao Wang, Jingmin Cheng, Tao Yang, Zhiyong Sun, Yongqin Kuang, Haidong Huang, Kexia Fan, Jianwen Gu
ABSTRACT
The prognosis of patients with malignant glioma is always quite poor, and this poor prognosis is probably due to our incomplete understanding of the molecular mechanisms underlying malignant glioma. It is known that myocyte enhancer factor-2D (MEF2D) plays an oncogenic role in hepatocellular carcinoma and promotes the survival of various types of cells. However, little is known about the expression profile and function of MEF2D in malignant glioma. In this study, we investigated the function and expression of MEF2D in malignant glioma. We found that in malignant glioma, there is an aberrantly high expression of MEF2D, which leads to poor prognosis of malignant glioma. The downregulation of MEF2D suppresses the proliferation of malignant glioma cell lines by inducing delay of S and G2/M phases of cell cycle and promoting apoptosis. Furthermore, the overexpression of MEF2D in astrocytes accelerates cell proliferation by regulating cell cycle progression. Furthermore, a mouse malignant glioma model demonstrated that MEF2D deficiency blocks malignant glioma formation in vivo. We conclude that MEF2D may act as a potential oncogene in malignant glioma and thus serve as a candidate target for malignant glioma therapy.
Sigma-Aldrich
Anti-MEF2D antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Roche
In Situ Cell Death Detection Kit, POD, sufficient for ≤50 tests