Merck
  • The RIF1-long splice variant promotes G1 phase 53BP1 nuclear bodies to protect against replication stress.
The RIF1-long splice variant promotes G1 phase 53BP1 nuclear bodies to protect against replication stress.
eLife (2020-11-04)
Lotte P Watts, Toyoaki Natsume, Yuichiro Saito, Javier Garzon, Qianqian Dong, Lora Boteva, Nick Gilbert, Masato T Kanemaki, Shin-Ichiro Hiraga, Anne D Donaldson
ABSTRACT
Human cells lacking RIF1 are highly sensitive to replication inhibitors, but the reasons for this sensitivity have been enigmatic. Here, we show that RIF1 must be present both during replication stress and in the ensuing recovery period to promote cell survival. Of two isoforms produced by alternative splicing, we find that RIF1-Long alone can protect cells against replication inhibition, but RIF1-Short is incapable of mediating protection. Consistent with this isoform-specific role, RIF1-Long is required to promote the formation of the 53BP1 nuclear bodies that protect unrepaired damage sites in the G1 phase following replication stress. Overall, our observations show that RIF1 is needed at several cell cycle stages after replication insult, with the RIF1-Long isoform playing a specific role during the ensuing G1 phase in damage site protection.
Sigma-Aldrich
Anti-DNA Antibody, single stranded, clone 16-19, clone 16-19, Chemicon®, from mouse
Sigma-Aldrich
Anti-PICH Antibody, clone 142-26-3, clone 142-26-3, from mouse