Merck
  • Citron Rho-Interacting Serine/Threonine Kinase Promotes HIF1a-CypA Signaling and Growth of Human Pancreatic Adenocarcinoma.
Citron Rho-Interacting Serine/Threonine Kinase Promotes HIF1a-CypA Signaling and Growth of Human Pancreatic Adenocarcinoma.
BioMed research international (2020-03-19)
Lin Cong, Zhile Bai, Yang Du, Yong Cheng
ABSTRACT
In human pancreatic ductal adenocarcinoma (PDAC), the cyclophilin A (CypA) is overexpressed and promotes the development of PDAC. However, the mechanism underlying cyclophilin A expression remains elusive. Here, we reported that the citron Rho-interacting serine/threonine kinase (CIT) promotes the HIF1a-CypA signaling and growth of PDAC cells. CIT expression was higher in PDAC cells compared with the normal epithelial cells, and clinical data showed that CIT was overexpressed in PDAC tissues and high expression of CIT predicted poor overall and disease-free survival. In PDAC cells, knockdown of CIT expression repressed the rate of proliferation and capacity of colony formation, which were accomplished with an increased percentage of apoptotic cells and cell cycle arrest. The knockdown of CIT in PDAC cells reduced the expression of CypA while overexpression of CIT promoted the expression of CypA. We observed that the effects of CIT on the expression of CypA relied on the transcriptional factor HIF1a, which was previously reported to transcriptionally activate the expression of CypA in PDAC cells. Furthermore, the effects of CIT on apoptosis, cell cycle, proliferation, and colony formation of PDAC cells relied on its role in the regulation of CypA expression. Collectively, our data showed that CIT promoted the activation of HIF1-CypA signaling and enhanced the growth of PDAC cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human PPIA
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder