Merck
  • Thrombin-cleaved osteopontin regulates hemopoietic stem and progenitor cell functions through interactions with alpha9beta1 and alpha4beta1 integrins.
Thrombin-cleaved osteopontin regulates hemopoietic stem and progenitor cell functions through interactions with alpha9beta1 and alpha4beta1 integrins.
Blood (2009-05-07)
Jochen Grassinger, David N Haylock, Melonie J Storan, Gemma O Haines, Brenda Williams, Genevieve A Whitty, Andrew R Vinson, Cheang Ly Be, Songhui Li, Esben S Sørensen, Patrick P L Tam, David T Denhardt, Dean Sheppard, Peter F Choong, Susan K Nilsson
ABSTRACT
Osteopontin (OPN), a multifunctional acidic glycoprotein, expressed by osteoblasts within the endosteal region of the bone marrow (BM) suppresses the proliferation of hemopoietic stem and progenitor cells and also regulates their lodgment within the BM after transplantation. Herein we demonstrate that OPN cleavage fragments are the most abundant forms of this protein within the BM. Studies aimed to determine how hemopoietic stem cells (HSCs) interact with OPN revealed for the first time that murine and human HSCs express alpha(9)beta(1) integrin. The N-terminal thrombin cleavage fragment of OPN through its binding to the alpha(9)beta(1) and alpha(4)beta(1) integrins plays a key role in the attraction, retention, regulation, and release of hemopoietic stem and progenitor cells to, in, and from their BM niche. Thrombin-cleaved OPN (trOPN) acts as a chemoattractant for stem and progenitor cells, mediating their migration in a manner that involves interaction with alpha(9)beta(1) and alpha(4)beta(1) integrins. In addition, in the absence of OPN, there is an increased number of white blood cells and, specifically, stem and progenitor cells in the peripheral circulation.
Sigma-Aldrich
Anti-Integrin α9β1 Antibody, clone Y9A2, azide free, clone Y9A2, Chemicon®, from mouse