Merck
Novel herbal medicine LA16001 ameliorates cisplatin-induced anorexia.
Molecular medicine reports (2017-12-06)
Sang-Mi Woo, Kang Min Lee, Gyu Ri Lee, Ji Youn Park, Hee Jae Lee, Hyo-Jung Bahn, Hyun Seok Yoon, Jem Yung Kim, Young Cheol Shin, Sung-Gook Cho, Seong-Gyu Ko
ABSTRACT
Chemotherapy frequently causes anorexia in cancer patients, which has been associated with poor disease prognosis. Several therapeutic strategies for the treatment of chemotherapy‑induced anorexia are available; however, their adverse effects limit their clinical use. Herbal medicines have a long history of use for the treatment of various diseases, including cancer, and recent research has demonstrated their safety and efficacy. In the present study, combinations of herbal medicines were designed based on traditional Korean medicine, and their effects were investigated on chemotherapy‑induced anorexia. Herbal mixtures were extracted, composed of Atractylodes japonica, Angelica gigas, Astragalus membranaceus, Lonicera japonica Thunb., Taraxacum platycarpum H. Dahlstedt and Prunella vulgaris var. asiatica (Nakai) Hara. The mixtures were termed LCBP‑Anocure‑16001‑3 (LA16001, LA16002, LA16003). A cisplatin‑induced anorexic mouse model was used to evaluate the putative effects of the extracts on chemotherapy‑induced anorexia. Treatment with LA16001 was revealed to prevent body weight loss, and all three extracts were demonstrated to improve food intake. When the molecular mechanisms underlying the orexigenic effects of LA16001 were investigated, altered expression levels of ghrelin, leptin and interleukin‑6 were revealed. Furthermore, LA16001 was reported to induce phosphorylation of Janus kinase 1 and signal transducer and activator of transcription 3. In addition, LA16001 administration increased the number of white blood cells and neutrophils. These results suggested that the herbal formula LA16001 may be able to prevent chemotherapy‑induced anorexia and may have potential as a novel therapeutic strategy for the adjuvant treatment of patients with cancer.
Product Description
Millipore
Rat/Mouse Ghrelin (active) ELISA, This Rat/Mouse Ghrelin (active) ELISA is used to measure & quantify Ghrelin levels in Metabolism & Endocrine research.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2