Merck
  • FMR1 Reactivating Treatments in Fragile X iPSC-Derived Neural Progenitors In Vitro and In Vivo.
FMR1 Reactivating Treatments in Fragile X iPSC-Derived Neural Progenitors In Vitro and In Vivo.
Cell reports (2019-03-07)
Dan Vershkov, Nina Fainstein, Sapir Suissa, Tamar Golan-Lev, Tamir Ben-Hur, Nissim Benvenisty
ABSTRACT
Fragile X syndrome (FXS) is caused primarily by a CGG repeat expansion in the FMR1 gene that triggers its transcriptional silencing. In order to investigate the regulatory layers involved in FMR1 inactivation, we tested a collection of chromatin modulators for the ability to reactivate the FMR1 locus. Although inhibitors of DNA methyltransferase (DNMT) induced the highest levels of FMR1 expression, a combination of a DNMT inhibitor and another compound potentiated the effect of reactivating treatment. To better assess the rescue effect following direct demethylation, we characterized the long-term and genome-wide effects of FMR1 reactivation and established an in vivo system to analyze FMR1-reactivating therapies. Systemic treatment with a DNMT inhibitor in mice carrying FXS induced pluripotent stem cell (iPSC)-derived transplants robustly induced FMR1 expression in the affected tissue, which was maintained for a prolonged period of time. Finally, we show a proof of principle for FMR1-reactivating therapy in the context of the CNS.
Sigma-Aldrich
Anti-Nestin Antibody, clone rat-401, clone rat-401, Chemicon®, from mouse
Sigma-Aldrich
Anti-Mitochondria Antibody, surface of intact mitochondria, clone 113-1, clone 113-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®