
Salbutamol Impurity K

Synonym(s):
2-(tert-Butylamino)-1-[3-chloro-4-hydroxy-5-(hydroxymethyl)phenyl]ethanone, 2-[(1,1-Dimethylethyl)amino]-1-[3-chloro-4-hydroxy-5-(hydroxymethyl)phenyl]ethanone
Empirical Formula (Hill Notation):
C13H18ClNO3
CAS Number:
Molecular Weight:
271.74
Products
Brand
Formula Weight
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing