
Paromomycin sulfate

Synonym(s):
Paromomycin sulfate salt, Aminosidine sulfate
Empirical Formula (Hill Notation):
C23H45N5O14 · xH2SO4
CAS Number:
Molecular Weight:
615.63 (free base basis)
Products
Product Category
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing