
N-Desmethyl U-47700 solution

Empirical Formula (Hill Notation):
C15H20Cl2N2O
CAS Number:
Molecular Weight:
315.24
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing