![Lithium acetoacetate analytical standard](/deepweb/assets/sigmaaldrich/product/structures/414/023/3b667178-bad9-409c-8511-3cd0079b5183/640/3b667178-bad9-409c-8511-3cd0079b5183.png)
Lithium acetoacetate
![Lithium acetoacetate analytical standard](/deepweb/assets/sigmaaldrich/product/structures/414/023/3b667178-bad9-409c-8511-3cd0079b5183/640/3b667178-bad9-409c-8511-3cd0079b5183.png)
Synonym(s):
Acetoacetic acid lithium salt
Linear Formula:
CH3COCH2COOLi
CAS Number:
Molecular Weight:
108.02
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing