Skip to Content
Merck
Klebsiella Selective Supplement suitable for microbiology

Klebsiella Selective Supplement

Klebsiella Selective Supplement suitable for microbiology
Synonym(s):
Carbenicillin, α-Carboxybenzylpenicillin solution
Empirical Formula (Hill Notation):
C17H18N2O6S
Molecular Weight:
378.40
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Application
Microorganism Suitability
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing