Skip to Content
Merck
Folic acid for impurity I identification European Pharmacopoeia (EP) Reference Standard

Folic acid for impurity I identification

Folic acid for impurity I identification European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Folic acid, PteGlu, Pteroyl-L-glutamic acid, Vitamin M
Empirical Formula (Hill Notation):
C19H19N7O6
CAS Number:
Molecular Weight:
441.40
EC Number:

Products

Product Category
Biological Source
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1